(1R,2R)-2-Aminocyclohexanecarboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A503601-100MG A503601-1G A503601-250MG
77156-646EA 128.75 USD
77156-646 77156-648 77156-650
(1R,2R)-2-Aminocyclohexanecarboxylic acid hydrochloride 95%
(1R,2R)-2-Aminocyclohexanecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MW: 179.64 g/mol
MDL Number: MFCD18374348
CAS Number: 158414-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR