5,6,7,8-Tetrahydronaphthalene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5,6,7,8-Tetrahydro-2-naphthoic acid

A502250-5G A502250-250MG
77156-644EA 116.39 USD
77156-644 77686-315
5,6,7,8-Tetrahydronaphthalene-2-carboxylic acid 98%
5,6,7,8-Tetrahydronaphthalene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD01861899
CAS Number: 1131-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR