7,8-Difluoroquinolin-3-amine 95%

Supplier: Ambeed
Warning

A501595-100MG A501595-1G A501595-250MG
77156-628EA 199.82 USD
77156-628 77156-630 77156-632
7,8-Difluoroquinolin-3-amine 95%
7,8-Difluoroquinolin-3-amine

Precaución: for Research Use Only

Formula: C₉H₆F₂N₂
MW: 180.15 g/mol
MDL Number: MFCD20723366
CAS Number: 318685-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR