Ethyl (3-hydroxycyclobutyl)acetate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(3-hydroxycyclobutyl)acetate

A499967-100MG A499967-1G A499967-250MG
77156-624EA 70 USD
77156-624 77686-310 77686-311
Ethyl (3-hydroxycyclobutyl)acetate 95%
Ethyl (3-hydroxycyclobutyl)acetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MDL Number: MFCD23105932
CAS Number: 1408075-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR