(3-Fluorobenzyl)hydrazine dihydrochloride 95%

Supplier: Ambeed
Warning

A495675-100MG A495675-1G A495675-250MG
77156-614EA 18 USD
77156-614 77156-616 77156-618
(3-Fluorobenzyl)hydrazine dihydrochloride 95%
(3-Fluorobenzyl)hydrazine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁Cl₂FN₂
MW: 213.08 g/mol
MDL Number: MFCD20233303
CAS Number: 1000805-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR