Potassium-3-fluoro-4-phenylphenyltrifluoroborate 96%

Supplier: Ambeed
Warning

Synonyms: Potassium (2-fluorobiphenyl-4-yl)trifluoroborate, Potassium trifluoro(2-fluorobiphenyl-4-yl)borate

A494217-1G A494217-250MG A494217-5G
77156-602EA 72 USD
77156-602 77156-604 77156-606
Potassium-3-fluoro-4-phenylphenyltrifluoroborate 96%
Potassium 3-fluoro-4-phenylphenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₁₂H₈BF₄K
MDL Number: MFCD08235035
CAS Number: 850623-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR