5-Ethyl-2-methoxybenzene-1-sulfonyl chloride 98%

Supplier: Ambeed
Danger

A494049-1G A494049-250MG A494049-5G
77156-594EA 50 USD
77156-594 77156-596 77156-598
5-Ethyl-2-methoxybenzene-1-sulfonyl chloride 98%
5-Ethyl-2-methoxybenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₃S
MDL Number: MFCD03424997
CAS Number: 88040-88-4
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR