Delamanid 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Methyl-6-nitro-2-((4-(4-(4-(trifluoromethoxy)phenoxy)piperidin-1-yl)phenoxy)methyl)-2,3-dihydroimidazo[2,1-b]oxazole

A493940-100MG A493940-1MG A493940-5MG A493940-250MG A493940-1G A493940-50MG A493940-5G
77156-588EA 185.4 USD
77156-588 77156-590 77156-592 77613-558 77686-293 77686-294 77686-295
Delamanid 95%
Delamanid

Precaución: for Research Use Only

Formula: C₂₅H₂₅F₃N₄O₆
MW: 534.492 g/mol
MDL Number: MFCD18251539
CAS Number: 681492-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR