(9aS)-Octahydropyrazino[2,1-c][1,4]oxazine 98%

Supplier: Ambeed
Warning

A493562-100MG A493562-1G A493562-250MG
77156-582EA 127.72 USD
77156-582 77156-584 77156-586
(9aS)-Octahydropyrazino[2,1-c][1,4]oxazine 98%
(9aS)-Octahydropyrazino[2,1-c][1,4]oxazine

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O
MW: 142.19 g/mol
MDL Number: MFCD22581549
CAS Number: 1089759-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR