3-Methyl-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

A491533-1G A491533-5G
77156-580EA 91.67 USD
77156-580 77613-552
3-Methyl-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one 98%
3-Methyl-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD18382389
CAS Number: 1330055-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR