Tributylmethylammonium bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Warning

A489490-1G A489490-250MG A489490-25G A489490-5G
77156-542EA 6 USD
77156-542 77156-544 77156-546 77156-548
Tributylmethylammonium bis(trifluoromethanesulfonyl)imide 98%
Tributylmethylammonium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₅H₃₀F₆N₂O₄S₂
MDL Number: MFCD12761447
CAS Number: 405514-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR