Methyl 5-chloropyrimidine-2-carboxylate 95%

Supplier: Ambeed
Warning

A488514-100MG A488514-250MG A488514-5G A488514-1G
77156-536EA 13 USD
77156-536 77156-538 77156-540 77901-432
Methyl 5-chloropyrimidine-2-carboxylate 95%
Methyl 5-chloropyrimidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD16611603
CAS Number: 894797-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR