3-Bromo-2-fluoro-5-methylbenzoic acid 95%

Supplier: Ambeed
Warning

A487343-1G A487343-25G A487343-5G A487343-250MG
77156-516EA 17 USD
77156-516 77156-518 77156-520 77686-277
3-Bromo-2-fluoro-5-methylbenzoic acid 95%
3-Bromo-2-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MDL Number: MFCD00506954
CAS Number: 72518-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR