(3-Bromo-4-chlorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-chlorophenylboronic acid

A486535-1G A486535-250MG A486535-5G
77156-502EA 175.1 USD
77156-502 77156-504 77156-506
(3-Bromo-4-chlorophenyl)boronic acid 98%
(3-Bromo-4-chlorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrClO₂
MW: 235.27 g/mol
MDL Number: MFCD19981540
CAS Number: 1384956-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR