trans-4-(Morpholin-4-yl)cyclohexylamine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: trans-4-Morpholinocyclohexanamine dihydrochloride, (1r,4r)-4-Morpholinocyclohexanamine dihydrochloride

A485951-100MG A485951-250MG A485951-1G A485951-5G
77156-490EA 46 USD
77156-490 77156-492 77686-276 77901-362
trans-4-(Morpholin-4-yl)cyclohexylamine dihydrochloride 95%
trans-4-(Morpholin-4-yl)cyclohexylamine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O·2ClH
MDL Number: MFCD27978481
CAS Number: 412356-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR