tert-Butyl 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine-7-carboxylate 98%

Supplier: Ambeed
Danger

A484087-100MG A484087-1G A484087-250MG A484087-5G
77156-482EA 20.6 USD
77156-482 77156-484 77156-486 77156-488
tert-Butyl 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine-7-carboxylate 98%
tert-Butyl 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClIN₃O₂
MW: 379.58 g/mol
MDL Number: MFCD20922795
CAS Number: 1244855-76-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR