Methyl (6-chloro-5-methyl-pyridazin-3-yl)acetate 95%

Supplier: Ambeed
Warning

A483707-1G A483707-250MG
77156-478EA 89 USD
77156-478 77156-480
Methyl (6-chloro-5-methyl-pyridazin-3-yl)acetate 95%
Methyl (6-chloro-5-methyl-pyridazin-3-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MDL Number: MFCD23106037
CAS Number: 1408074-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR