Propargyl-PEG1-NHS ester 98%

Supplier: Ambeed
Warning

Synonyms: N-Succinimidyl 3-(propargyloxy)propionate, Propargyl-dPEG®1-NHS ester

A483542-1G A483542-50MG A483542-100MG
77156-476EA 268 USD
77156-476 77568-950 77686-274
Propargyl-PEG1-NHS ester 98%
Propargyl-PEG1-NHS ester

Precaución: For research use only.

Formula: C₁₀H₁₁NO₅
MW: 225.2 g/mol
MDL Number: MFCD13184973
CAS Number: 1174157-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR