5-Amino-6-chloro-2,3-dicyanopyrazine 98%

Supplier: Ambeed
Danger

Synonyms: 5-Amino-6-chloro-2,3-pyrazinedicarbonitrile, 5-Amino-6-chloropyrazine-2,3-dicarbonitrile

A483408-1G A483408-25G A483408-5G
77156-474EA 30.9 USD
77156-474 77551-468 77572-526
5-Amino-6-chloro-2,3-dicyanopyrazine 98%
5-Amino-6-chloro-2,3-dicyanopyrazine

Precaución: for Research Use Only

Formula: C₆H₂ClN₅
MW: 179.57 g/mol
MDL Number: MFCD00191417
CAS Number: 56413-96-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR