Ethyl 5-(4-fluorophenyl)-5-oxovalerate 98%

Supplier: Ambeed
Warning

A482767-1G A482767-250MG A482767-5G
77156-468EA 7 USD
77156-468 77156-470 77156-472
Ethyl 5-(4-fluorophenyl)-5-oxovalerate 98%
Ethyl 5-(4-fluorophenyl)-5-oxovalerate

Precaución: for Research Use Only

Formula: C₁₃H₁₅FO₃
MDL Number: MFCD01311503
CAS Number: 342636-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR