tert-Butyl 4-acryloylpiperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A481408-100MG A481408-1G A481408-250MG
77156-460EA 87.55 USD
77156-460 77156-462 77156-464
tert-Butyl 4-acryloylpiperazine-1-carboxylate 95%
tert-Butyl 4-acryloylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₃
MW: 240.3 g/mol
MDL Number: MFCD24470187
CAS Number: 270594-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR