1-Naphthol 97%

Supplier: Ambeed
Danger

A480985-100G A480985-25G A480985-500G
77156-454EA 47 USD
77156-454 77156-456 77156-458
1-Naphthol 97%
1-Naphthol

Precaución: for Research Use Only

Formula: C₁₀H₈O
MDL Number: MFCD00003930
CAS Number: 90-15-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR