5-Fluoro-2-(trifluoromethoxy)benzoic acid 98%

Supplier: Ambeed
Warning

A480842-100MG A480842-1G A480842-250MG
77156-448EA 18.54 USD
77156-448 77156-450 77156-452
5-Fluoro-2-(trifluoromethoxy)benzoic acid 98%
5-Fluoro-2-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
MDL Number: MFCD11519333
CAS Number: 1092460-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR