trans-1-(Boc-amino)-4-ethynylcyclohexane 95%

Supplier: Ambeed
Warning

A479658-100MG A479658-1G A479658-250MG A479658-5G
77156-414EA 52 USD
77156-414 77156-416 77156-418 77156-420
trans-1-(Boc-amino)-4-ethynylcyclohexane 95%
trans-1-(Boc-amino)-4-ethynylcyclohexane

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₂
MDL Number: MFCD12963985
CAS Number: 947141-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR