Acridine orange (hemizinc salt) for electrophoresis

Supplier: Ambeed
Danger

Synonyms: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

A479198-250MG A479198-1G A479198-5G A479198-25G
77156-398EA 6 USD
77156-398 77613-496 77613-498 77686-265
Acridine orange (hemizinc salt) for electrophoresis
Acridine orange (hemizinc salt)

Precaución: for Research Use Only

Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MW: 369.96 g/mol
MDL Number: MFCD00081043
CAS Number: 10127-02-3
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR