Ethyl 6-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A476855-100G
A476855-25G
A476855-5G
77156-382EA
552.08
USD
77156-382
77156-384
77156-386
Ethyl 6-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylate 98%
Ethyl 6-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₀BrNO₃ MW: 296.11 g/mol |
MDL Number:
MFCD00173362 CAS Number: 79607-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...