Methyl-4-chloro-3-iodobenzoate 98%

Supplier: Ambeed
Warning

A475932-5G A475932-25G
77156-380EA 39 USD
77156-380 77901-180
Methyl-4-chloro-3-iodobenzoate 98%
Methyl 4-chloro-3-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClIO₂
MDL Number: MFCD09263357
CAS Number: 365526-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR