(R)-1-Boc-4,4-Difluoropyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A472492-1G A472492-250MG A472492-5G A472492-100MG
77156-348EA 87 USD
77156-348 77156-350 77686-254 77892-956
(R)-1-Boc-4,4-Difluoropyrrolidine-2-carboxylic acid 97%
(R)-1-Boc-4,4-Difluoropyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅F₂NO₄
MDL Number: MFCD08272893
CAS Number: 536747-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR