tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A471786-100MG A471786-1G A471786-250MG A471786-5G
77156-334EA 19.57 USD
77156-334 77156-336 77156-338 77156-340
tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate 98%
tert-Butyl 4-hydroxy-2-methylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.289 g/mol
MDL Number: MFCD16497040
CAS Number: 208046-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR