tert-Butyl 3-(cyanomethyl)piperidine-1-carboxylate 96%

Supplier: Ambeed
Warning

A469801-1G A469801-250MG A469801-5G
77156-304EA 73 USD
77156-304 77156-306 77901-586
tert-Butyl 3-(cyanomethyl)piperidine-1-carboxylate 96%
tert-Butyl 3-(cyanomethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₂
MDL Number: MFCD08461079
CAS Number: 948015-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR