N-(3-Chlorophenyl)picolinamide 99+%

Supplier: Ambeed
Warning

Synonyms: VU0364770

A469094-100MG A469094-10MG A469094-1MG A469094-25MG A469094-50MG A469094-5MG A469094-250MG
77156-284EA 202.91 USD
77156-284 77156-286 77156-288 77156-290 77156-292 77156-294 77686-249
N-(3-Chlorophenyl)picolinamide 99+%
N-(3-Chlorophenyl)picolinamide

Precaución: for Research Use Only

Formula: C₁₂H₉ClN₂O
MW: 232.66 g/mol
MDL Number: MFCD00548412
CAS Number: 61350-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR