T0901317 95%

Supplier: Ambeed
Warning

Synonyms: N-(4-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenyl)-N-(2,2,2-trifluoroethyl)benzenesulphonamide

A467377-10MG A467377-1MG A467377-50MG A467377-5MG A467377-1G A467377-250MG A467377-100MG
77156-276EA 61.8 USD
77156-276 77156-278 77156-280 77156-282 77613-442 77613-444 77686-244
T0901317 95%
T0901317

Precaución: for Research Use Only

Formula: C₁₇H₁₂F₉NO₃S
MW: 481.3327 g/mol
MDL Number: MFCD03412028
CAS Number: 293754-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR