4,8-Dimethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,6,2-dioxazaborocane 95+%

Supplier: Ambeed
Warning

A467309-1G A467309-250MG A467309-5G
77156-270EA 202.91 USD
77156-270 77156-272 77156-274
4,8-Dimethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,6,2-dioxazaborocane 95+%
4,8-Dimethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,6,2-dioxazaborocane

Precaución: for Research Use Only

Formula: C₁₂H₂₅B₂NO₄
MW: 268.95 g/mol
MDL Number: MFCD28015459
CAS Number: 1536530-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR