tert-Butyl 3-bromo-1H-pyrrolo[3,2-b]pyridine-1-carboxylate 98+%

Supplier: Ambeed
Warning

A465577-1G A465577-250MG
77156-262EA 97.85 USD
77156-262 77156-264
tert-Butyl 3-bromo-1H-pyrrolo[3,2-b]pyridine-1-carboxylate 98+%
tert-Butyl 3-bromo-1H-pyrrolo[3,2-b]pyridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O₂
MW: 297.14 g/mol
MDL Number: MFCD16660027
CAS Number: 192189-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR