3-Acetylpyridine N-Oxide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Acetylpyridine n-oxide

A464864-1G A464864-250MG A464864-5G
77156-252EA 75.19 USD
77156-252 77156-254 77156-256
3-Acetylpyridine N-Oxide 98%
3-Acetylpyridine N-Oxide

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00955798
CAS Number: 14188-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR