Methyl (1S,3R,4aS,8S,8aS)-3-hydroxy-1-methyl-8-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4,4a,8,8a-tetrahydro-1H,3H-pyrano[3,4-c]pyran-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Morroniside

A464826-10MG A464826-1MG A464826-25MG A464826-50MG
77156-244EA 42.23 USD
77156-244 77156-246 77156-248 77156-250
Methyl (1S,3R,4aS,8S,8aS)-3-hydroxy-1-methyl-8-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4,4a,8,8a-tetrahydro-1H,3H-pyrano[3,4-c]pyran-5-carboxylate 95%
Methyl (1S,3R,4aS,8S,8aS)-3-hydroxy-1-methyl-8-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4,4a,8,8a-tetrahydro-1H,3H-pyrano[3,4-c]pyran-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₆O₁₁
MW: 406.38 g/mol
MDL Number: MFCD00210532
CAS Number: 25406-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR