3-(Dimethylamino)-1-(2-thienyl)-1-propanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene

A463583-100G A463583-1G A463583-25G A463583-5G
77156-236EA 157.59 USD
77156-236 77156-238 77156-240 77156-242
3-(Dimethylamino)-1-(2-thienyl)-1-propanol 98%
3-(Dimethylamino)-1-(2-thienyl)-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₅NOS
MW: 185.29 g/mol
MDL Number: MFCD09701963
CAS Number: 13636-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR