4'-Methyl-3'-nitroacetanilide 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-4-nitro-N-acetylbenzeneamine, N-(4-Methyl-3-nitrophenyl)acetamide

A463237-1G A463237-25G A463237-5G
77156-224EA 10.3 USD
77156-224 77156-226 77156-228
4'-Methyl-3'-nitroacetanilide 95%
4'-Methyl-3'-nitroacetanilide

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₃
MW: 194.187 g/mol
MDL Number: MFCD00024277
CAS Number: 2719-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR