2-(Trimethylsilyloxy)ethyl methacrylate 96% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with BHT or MEHQ

A462971-25G A462971-5G A462971-100G
77156-220EA 42 USD
77156-220 77156-222 77572-460
2-(Trimethylsilyloxy)ethyl methacrylate 96% stabilized
2-(Trimethylsilyloxy)ethyl methacrylate

Precaución: for Research Use Only

Formula: C₉H₁₈O₃Si
MDL Number: MFCD00053869
CAS Number: 17407-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR