2-Bromo-5-(cyclopropylmethoxy)pyridine 98%

Supplier: Ambeed
Warning

A462684-1G A462684-5G
77156-214EA 104.03 USD
77156-214 77156-216
2-Bromo-5-(cyclopropylmethoxy)pyridine 98%
2-Bromo-5-(cyclopropylmethoxy)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MW: 228.08 g/mol
MDL Number: MFCD22690535
CAS Number: 1177269-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR