2,4,6-Trichlorophenylboronic acid 98%

Supplier: Ambeed
Warning

A458925-10G A458925-1G A458925-250MG A458925-25G A458925-5G
77156-176EA 35 USD
77156-176 77156-178 77156-180 77156-182 77156-184
2,4,6-Trichlorophenylboronic acid 98%
2,4,6-Trichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₃O₂
MDL Number: MFCD01074621
CAS Number: 73852-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR