4-Fluoro-3,5-dimethylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoro-3,5-dimethylphenyl)methanol

A458834-1G A458834-250MG
77156-172EA 71.07 USD
77156-172 77156-174
4-Fluoro-3,5-dimethylbenzyl alcohol 95%
4-Fluoro-3,5-dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
MDL Number: MFCD06660267
CAS Number: 886501-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR