3,4,6-Tri-O-benzyl-D-galactal 98%

Supplier: Ambeed
Warning

Synonyms: Tri-O-Benzyl-D-galactal

A454727-1G A454727-5G
77156-156EA 47.38 USD
77156-156 77156-158
3,4,6-Tri-O-benzyl-D-galactal 98%
3,4,6-Tri-O-benzyl-D-galactal

Precaución: for Research Use Only

Formula: C₂₇H₂₈O₄
MW: 416.517 g/mol
MDL Number: MFCD09039276
CAS Number: 80040-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR