1-Indan-2-ylethanone 95%

Supplier: Ambeed
Warning

A451661-1G A451661-5G
77156-134EA 11 USD
77156-134 77156-136
1-Indan-2-ylethanone 95%
1-Indan-2-ylethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MDL Number: MFCD08543446
CAS Number: 33982-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR