3-Bromoquinoline-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A450799-100MG A450799-250MG A450799-5G
77156-132EA 23.69 USD
77156-132 77686-215 77686-216
3-Bromoquinoline-6-carboxylic acid 95%
3-Bromoquinoline-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MW: 252.064 g/mol
MDL Number: MFCD18086942
CAS Number: 205114-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR