Cyclohexylhydrazine hydrochloride 97%

Supplier: Ambeed
Warning

A449845-100G A449845-10G A449845-1G A449845-5G A449845-25G
77156-124EA 410.01 USD
77156-124 77156-126 77156-128 77156-130 77568-886
Cyclohexylhydrazine hydrochloride 97%
Cyclohexylhydrazine hydrochloride

Precaución: For research use only.

Formula: C₆H₁₄N₂
MW: 150.65 g/mol
MDL Number: MFCD00060160
CAS Number: 24214-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR