N,N-Diethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1-sulfonamide 95+%

Supplier: Ambeed
Warning

A449750-1G A449750-250MG A449750-5G
77156-116EA 196.73 USD
77156-116 77156-118 77156-120
N,N-Diethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1-sulfonamide 95+%
N,N-Diethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1-sulfonamide

Precaución: for Research Use Only

Formula: C₁₆H₂₆BNO₄S
MW: 339.258 g/mol
MDL Number: MFCD27957314
CAS Number: 911228-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR