(Oxybis(4,1-phenylene))bis(dimethylsilane) 97%

Supplier: Ambeed
Danger

A448209-1G A448209-250MG
77156-110EA 16.48 USD
77156-110 77156-112
(Oxybis(4,1-phenylene))bis(dimethylsilane) 97%
(Oxybis(4,1-phenylene))bis(dimethylsilane)

Precaución: for Research Use Only

Formula: C₁₆H₂₂OSi₂
MW: 286.52 g/mol
MDL Number: MFCD00054791
CAS Number: 13315-17-8
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR