3,6-Dihydro-2H-pyran-4-carbonitrile 97%

Supplier: Ambeed
Danger

A447368-100MG A447368-250MG
77156-108EA 36.05 USD
77156-108 77686-211
3,6-Dihydro-2H-pyran-4-carbonitrile 97%
3,6-Dihydro-2H-pyran-4-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.126 g/mol
MDL Number: MFCD20040386
CAS Number: 105772-13-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR