1-Butyl-3-methylimidazolium methanesulfonate 98%

Supplier: Ambeed
Danger

A442178-1G A442178-25G A442178-5G
77156-068EA 13.39 USD
77156-068 77156-070 77156-072
1-Butyl-3-methylimidazolium methanesulfonate 98%
1-Butyl-3-methylimidazolium methanesulphonate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₃S
MDL Number: MFCD06798173
CAS Number: 342789-81-5
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR